Assay Details
| Gene Symbol: Csrnp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108786.1 | NP_001102256.1 | 1 - 2 | 68 | 79 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108786.1 transcripts.
Set Membership: |
|

Csrnp1
Genomic Map