Assay Details
| Gene Symbol: Ucn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133385.2 | NP_596876.2 | 2 | 268 | 56 | |
| GenBank mRNA | AY044835.1 | - | 1 | 244 | 56 | |
| EF078966.1 | - | 2 | 268 | 56 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ucn2
Genomic Map