Assay Details
| Gene Symbol: Hemk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106853.1 | NP_001100323.1 | 8 - 9 | 959 | 66 | |
| XM_006243728.2 | XP_006243790.1 | 9 - 10 | 1249 | 66 | ||
| XM_006243729.2 | XP_006243791.1 | 9 - 10 | 1569 | 66 | ||
| XM_006243730.2 | XP_006243792.1 | 8 - 9 | 1324 | 66 | ||
| XM_008766502.1 | XP_008764724.1 | 9 - 10 | 1184 | 66 | ||
| GenBank mRNA | BC161944.1 | - | 8 - 9 | 953 | 9024451 | 66 |
More Information
Updated Mapping Information
Set Membership: |
|

Hemk1
Genomic Map