Assay Details
| Gene Symbol: Higd1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080902.4 | NP_543178.2 | 1 - 2 | 58 | 73 | |
| GenBank mRNA | BC059118.1 | - | 1 - 2 | 62 | 6890532 | 73 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC059118.1,NM_080902.4 transcripts.
This assay is in the 5'UTR region of the BC059118.1,NM_080902.4 transcripts.
Set Membership: |
|

Higd1a
Genomic Map