Assay Details
| Gene Symbol: Cyb5r4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133427.3 | NP_596918.3 | 12 - 13 | 1246 | 63 | |
| XM_006243472.2 | XP_006243534.1 | 11 - 12 | 1232 | 63 | ||
| XM_008766437.1 | XP_008764659.1 | 12 - 13 | 1405 | 63 | ||
| GenBank mRNA | AF307840.1 | - | 11 - 12 | 1005 | 63 | |
| AY321370.1 | - | 11 - 12 | 1005 | 63 | ||
| BC080240.1 | - | 12 - 13 | 1224 | 7099950 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cyb5r4
Genomic Map