Assay Details
| Gene Symbol: Lrrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014268.1 | NP_001014290.1 | 13 - 14 | 1520 | 140 | |
| GenBank mRNA | BC079423.1 | - | 13 - 14 | 1520 | 7114094 | 140 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P448234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559085 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373618 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1554 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933417 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428849 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78175 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164816 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497045 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136754 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186674 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629272 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107089 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88195 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654854 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483370 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304451 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724902 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815877 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793374 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344012 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70818 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503852 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539122 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520874 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113600 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108381 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161640 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P448234 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P448234 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P448234 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P448234 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1032623 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 81352 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 57732 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 86395 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 14779 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 71639 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 26148 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 29970 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 84070 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35098 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 458 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 19477 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10918796 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918799 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918795 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918804 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918794 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918798 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918797 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918803 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918793 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918801 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918800 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918792 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918805 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918802 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10918791 | - | - |
| Rat Genome 230 2.0 Array | 1392901_at | - | - |
| Rat Genome 230 2.0 Array | 1385632_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1385632_at | - | - |
| Rat Genome 230 2.0 Array | 1392901_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1392901_at | - | - |
| Affymetrix HT Rat Focus Array | 1385632_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1392901_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1385632_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800023774.rn.1 | TC0800002969.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800023775.rn.1 | TC0800002969.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800023776.rn.1 | TC0800002969.rn.1 | RTA TC |
| Rat Expression Set 230 | 1392901_PM_at | - | - |
| Rat Expression Set 230 | 1385632_PM_at | - | - |
| Rat Expression Set 230 | 1392901_at | - | - |
| Rat Expression Set 230 | 1385632_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918791 | - | - |
| Rat Genome U34 Set | rc_AA925938_at | - | - |
| Rat Genome U34 Set | rc_AI137106_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22375570 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358368 | - | - |

Lrrc1
Genomic Map