Assay Details
| Gene Symbol: Faim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006243616.2 | XP_006243678.1 | 153 | 84 | ||
| Gene Symbol: LOC100362113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729959.3 | XP_002730005.1 | 1 - 2 | 153 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002729959.3, XM_006243616.2 transcripts.
Set Membership: |
|

Faim
Genomic Map