Assay Details
| Gene Symbol: Exog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106866.1 | NP_001100336.1 | 3 - 4 | 498 | 76 | |
| XM_006244088.2 | XP_006244150.1 | 4 - 5 | 559 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Exog
Genomic Map