Assay Details
| Gene Symbol: Manf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108183.1 | NP_001101653.1 | 1 - 2 | 143 | 94 | |
| XM_006243775.1 | XP_006243837.1 | 1 - 2 | 138 | 94 | ||
| GenBank mRNA | BC166980.1 | - | 1 - 2 | 115 | 7318164 | 94 |
| FQ232064.1 | - | 1 - 2 | 273 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Manf
Genomic Map