Assay Details
| Gene Symbol: Cmtm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007802.3 | NP_001007803.1 | 2 - 3 | 184 | 74 | |
| XM_006243969.2 | XP_006244031.1 | 1 - 2 | 206 | 74 | ||
| GenBank mRNA | AY325262.1 | - | 2 - 3 | 184 | 74 | |
| AY387088.1 | - | 1 - 2 | 144 | 74 | ||
| BC107930.1 | - | 1 - 2 | 177 | 7130158 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cmtm6
Genomic Map