Assay Details
| Gene Symbol: LOC100912347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006243414.2 | XP_006243476.1 | 4 - 5 | 2205 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100912347| Gene Symbol: LOC100912347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006243414.2 | XP_006243476.1 | 4 - 5 | 2205 | 112 | |
Set Membership: |
|