Assay Details
Gene Symbol: Mrap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108774.1 | NP_001102244.1 | 1 - 2 | 66 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108774.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
