Assay Details
| Gene Symbol: Mrap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108774.1 | NP_001102244.1 | 2 - 3 | 183 | 131 | |
| XM_006243487.2 | XP_006243549.1 | 3 - 4 | 463 | 131 | ||
| XM_006243488.2 | XP_006243550.1 | 2 - 3 | 318 | 131 | ||
| XM_008766452.1 | XP_008764674.1 | 3 - 4 | 451 | 131 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108774.1, XM_006243487.2, XM_006243488.2, XM_008766452.1 transcripts.
Set Membership: |
|

Mrap2
Genomic Map