Assay Details
| Gene Symbol: LOC100910021 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750567.3 | XP_003750615.1 | 3 - 4 | 816 | 130 | |
| Gene Symbol: Pik3cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006243642.2 | XP_006243704.1 | 20 - 21 | 2957 | 130 | |
| XM_008766567.1 | XP_008764789.1 | 15 - 16 | 2344 | 130 | ||
| GenBank mRNA | AJ012482.1 | - | 20 - 21 | 2940 | 130 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100910021
Genomic Map