Assay Details
| Gene Symbol: Camp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100724.1 | NP_001094194.1 | 2 - 3 | 307 | 66 | |
| GenBank mRNA | AF484553.1 | - | 2 - 3 | 292 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Camp
Genomic Map