Assay Details
| Gene Symbol: Cep162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277060.1 | NP_001263989.1 | 21 - 22 | 2975 | 68 | |
| XM_006243475.2 | XP_006243537.1 | 22 - 23 | 3309 | 68 | ||
| XM_006243476.2 | XP_006243538.1 | 21 - 22 | 3117 | 68 | ||
| XM_008766441.1 | XP_008764663.1 | 17 - 18 | 2443 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cep162
Genomic Map