Assay Details
| Gene Symbol: Stt3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170539.1 | NP_001164010.1 | 10 - 11 | 1599 | 63 | |
| GenBank mRNA | BC166740.1 | - | 10 - 11 | 1599 | 7937421 | 63 |
| FQ210494.1 | - | 10 - 11 | 1600 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stt3b
Genomic Map