Assay Details
| Gene Symbol: LOC102554812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006243891.2 | XP_006243953.1 | 1 - 2 | 570 | 87 | |
| Gene Symbol: Dhx30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013249.1 | NP_001013267.1 | 1 - 2 | 54 | 87 | |
| GenBank mRNA | BC091359.1 | - | 1 - 2 | 54 | 7318733 | 87 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC091359.1,NM_001013249.1 transcripts.
Set Membership: |
|

LOC102554812
Genomic Map