Assay Details
| Gene Symbol: Abhd14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007664.1 | NP_001007665.1 | 1 - 2 | 62 | 78 | |
| GenBank mRNA | BC076385.1 | - | 1 - 2 | 62 | 7108802 | 78 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC076385.1,NM_001007664.1 transcripts.
This assay is in the 5'UTR region of the BC076385.1,NM_001007664.1 transcripts.
Set Membership: |
|

Abhd14b
Genomic Map