Assay Details
| Gene Symbol: Abhd14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007664.1 | NP_001007665.1 | 2 - 3 | 284 | 70 | |
| XM_006243724.1 | XP_006243786.1 | 1 - 2 | 225 | 70 | ||
| GenBank mRNA | BC076385.1 | - | 2 - 3 | 284 | 7108802 | 70 |
More Information
Updated Mapping Information
Set Membership: |
|

Abhd14b
Genomic Map