Assay Details
| Gene Symbol: Rpsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008766672.1 | XP_008764894.1 | 1 - 2 | 101 | 81 | |
| GenBank mRNA | BC060578.1 | - | 1 - 2 | 46 | 6919235 | 81 |
| FQ209463.1 | - | 1 - 2 | 42 | 81 | ||
| FQ209469.1 | - | 1 - 2 | 42 | 81 | ||
| FQ209490.1 | - | 1 - 2 | 43 | 81 | ||
| FQ209729.1 | - | 1 - 2 | 42 | 81 | ||
| FQ211189.1 | - | 1 - 2 | 42 | 81 | ||
| FQ212660.1 | - | 1 - 2 | 42 | 81 | ||
| FQ212893.1 | - | 1 - 2 | 42 | 81 | ||
| FQ213275.1 | - | 1 - 2 | 42 | 81 | ||
| FQ213317.1 | - | 1 - 2 | 42 | 81 | ||
| FQ213334.1 | - | 1 - 2 | 42 | 81 | ||
| FQ213623.1 | - | 1 - 2 | 45 | 81 | ||
| FQ213835.1 | - | 1 - 2 | 43 | 81 | ||
| FQ213894.1 | - | 1 - 2 | 42 | 81 | ||
| FQ215242.1 | - | 1 - 2 | 42 | 81 | ||
| FQ215292.1 | - | 1 - 2 | 43 | 81 | ||
| FQ216063.1 | - | 1 - 2 | 42 | 81 | ||
| FQ216085.1 | - | 1 - 2 | 42 | 81 | ||
| FQ216130.1 | - | 1 - 2 | 42 | 81 | ||
| FQ216245.1 | - | 1 - 2 | 42 | 81 | ||
| FQ216788.1 | - | 1 - 2 | 44 | 81 | ||
| FQ216997.1 | - | 1 - 2 | 42 | 81 | ||
| FQ217664.1 | - | 1 - 2 | 42 | 81 | ||
| FQ219586.1 | - | 1 - 2 | 42 | 81 | ||
| FQ219785.1 | - | 1 - 2 | 45 | 81 | ||
| FQ220012.1 | - | 1 - 2 | 42 | 81 | ||
| FQ220017.1 | - | 1 - 2 | 40 | 81 | ||
| FQ220084.1 | - | 1 - 2 | 45 | 81 | ||
| FQ220126.1 | - | 1 - 2 | 42 | 81 | ||
| FQ220171.1 | - | 1 - 2 | 41 | 81 | ||
| FQ220184.1 | - | 1 - 2 | 42 | 81 | ||
| FQ220347.1 | - | 1 - 2 | 44 | 81 | ||
| FQ220468.1 | - | 1 - 2 | 42 | 81 | ||
| FQ220661.1 | - | 1 - 2 | 45 | 81 | ||
| FQ220822.1 | - | 1 - 2 | 42 | 81 | ||
| FQ220964.1 | - | 1 - 2 | 42 | 81 | ||
| FQ221103.1 | - | 1 - 2 | 42 | 81 | ||
| FQ221334.1 | - | 1 - 2 | 62 | 81 | ||
| FQ221443.1 | - | 1 - 2 | 43 | 81 | ||
| FQ221672.1 | - | 1 - 2 | 42 | 81 | ||
| FQ222093.1 | - | 1 - 2 | 44 | 81 | ||
| FQ222280.1 | - | 1 - 2 | 42 | 81 | ||
| FQ222542.1 | - | 1 - 2 | 44 | 81 | ||
| FQ222757.1 | - | 1 - 2 | 40 | 81 | ||
| FQ223223.1 | - | 1 - 2 | 42 | 81 | ||
| FQ225510.1 | - | 1 - 2 | 43 | 81 | ||
| FQ226054.1 | - | 1 - 2 | 42 | 81 | ||
| FQ226408.1 | - | 1 - 2 | 42 | 81 | ||
| FQ227056.1 | - | 1 - 2 | 42 | 81 | ||
| FQ227293.1 | - | 1 - 2 | 42 | 81 | ||
| FQ227600.1 | - | 1 - 2 | 43 | 81 | ||
| FQ228002.1 | - | 1 - 2 | 42 | 81 | ||
| FQ228212.1 | - | 1 - 2 | 43 | 81 | ||
| FQ229290.1 | - | 1 - 2 | 44 | 81 | ||
| FQ229305.1 | - | 1 - 2 | 44 | 81 | ||
| FQ229367.1 | - | 1 - 2 | 43 | 81 | ||
| FQ229516.1 | - | 1 - 2 | 42 | 81 | ||
| FQ229579.1 | - | 1 - 2 | 42 | 81 | ||
| FQ229598.1 | - | 1 - 2 | 43 | 81 | ||
| FQ229712.1 | - | 1 - 2 | 42 | 81 | ||
| FQ229787.1 | - | 1 - 2 | 42 | 81 | ||
| FQ229850.1 | - | 1 - 2 | 44 | 81 | ||
| FQ229859.1 | - | 1 - 2 | 40 | 81 | ||
| FQ229890.1 | - | 1 - 2 | 42 | 81 | ||
| FQ229918.1 | - | 1 - 2 | 62 | 81 | ||
| FQ229945.1 | - | 1 - 2 | 42 | 81 | ||
| FQ229970.1 | - | 1 - 2 | 42 | 81 | ||
| FQ230088.1 | - | 1 - 2 | 42 | 81 | ||
| FQ230106.1 | - | 1 - 2 | 42 | 81 | ||
| FQ230180.1 | - | 1 - 2 | 43 | 81 | ||
| FQ230258.1 | - | 1 - 2 | 42 | 81 | ||
| FQ231622.1 | - | 1 - 2 | 40 | 81 | ||
| FQ232305.1 | - | 1 - 2 | 44 | 81 | ||
| FQ232568.1 | - | 1 - 2 | 43 | 81 | ||
| FQ233068.1 | - | 1 - 2 | 42 | 81 | ||
| FQ233112.1 | - | 1 - 2 | 42 | 81 | ||
| FQ233340.1 | - | 1 - 2 | 44 | 81 | ||
| FQ233547.1 | - | 1 - 2 | 43 | 81 | ||
| FQ233919.1 | - | 1 - 2 | 42 | 81 | ||
| FQ234511.1 | - | 1 - 2 | 42 | 81 | ||
| FQ234742.1 | - | 1 - 2 | 42 | 81 | ||
| FQ234772.1 | - | 1 - 2 | 42 | 81 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC060578.1,XM_008766672.1 transcripts.
This assay is in the 5'UTR region of the BC060578.1,XM_008766672.1 transcripts.
Set Membership: |
|

Rpsa
Genomic Map