Assay Details
| Gene Symbol: Zbtb38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006243579.2 | XP_006243641.1 | 2 - 3 | 581 | 99 | |
| XM_006243580.2 | XP_006243642.1 | 4 - 5 | 707 | 99 | ||
| XM_006243582.2 | XP_006243644.1 | 2 - 3 | 510 | 99 | ||
| XM_006243584.1 | XP_006243646.1 | 2 - 3 | 164 | 99 | ||
| XM_008766521.1 | XP_008764743.1 | 5 - 6 | 866 | 99 | ||
| XM_008766522.1 | XP_008764744.1 | 6 - 7 | 877 | 99 | ||
| XM_008766524.1 | XP_008764746.1 | 3 - 4 | 641 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006243579.2, XM_006243580.2, XM_006243582.2, XM_006243584.1, XM_008766521.1, XM_008766522.1, XM_008766524.1 transcripts.
Set Membership: |
|

Zbtb38
Genomic Map