Assay Details
| Gene Symbol: Rbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012733.4 | NP_036865.1 | 2 - 3 | 347 | 77 | |
| GenBank mRNA | FQ209770.1 | - | 2 - 3 | 361 | 77 | |
| FQ209927.1 | - | 2 - 3 | 331 | 77 | ||
| FQ210835.1 | - | 2 - 3 | 362 | 77 | ||
| FQ218090.1 | - | 2 - 3 | 361 | 77 | ||
| M16459.1 | - | 2 - 3 | 347 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbp1
Genomic Map