Assay Details
| Gene Symbol: Abhd14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009670.1 | NP_001009670.1 | 2 - 3 | 503 | 62 | |
| XM_006243717.2 | XP_006243779.1 | 1 - 2 | 344 | 62 | ||
| XM_008766494.1 | XP_008764716.1 | 2 - 3 | 915 | 62 | ||
| XM_008766495.1 | XP_008764717.1 | 3 - 4 | 569 | 62 | ||
| XM_008766496.1 | XP_008764718.1 | 2 - 3 | 518 | 62 | ||
| XM_008766497.1 | XP_008764719.1 | 2 - 3 | 387 | 62 | ||
| XM_008766498.1 | XP_008764720.1 | 1 - 2 | 339 | 62 | ||
| XM_008766499.1 | XP_008764721.1 | 1 - 2 | 344 | 62 | ||
| GenBank mRNA | BC088472.1 | - | 2 - 3 | 503 | 7128506 | 62 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC088472.1,NM_001009670.1,XM_006243717.2,XM_008766494.1,XM_008766495.1,XM_008766496.1,XM_008766497.1,XM_008766498.1,XM_008766499.1 transcripts.
Set Membership: |
|

Abhd14a
Genomic Map