Assay Details
| Gene Symbol: Cish | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031804.1 | NP_113992.1 | 1 - 2 | 135 | 71 | |
| GenBank mRNA | BC161930.1 | - | 1 - 2 | 135 | 8375557 | 71 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC161930.1,NM_031804.1 transcripts.
Set Membership: |
|

Cish
Genomic Map