Assay Details
| Gene Symbol: Nck1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106851.2 | NP_001100321.1 | 3 - 4 | 1078 | 115 | |
| GenBank mRNA | BC167009.1 | - | 3 - 4 | 1078 | 9034703 | 115 |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of protein kinase activity
substrate-dependent cell migration, cell extension
actin filament organization
cell migration
lamellipodium assembly
positive regulation of actin filament polymerization
positive regulation of translation in response to stress
negative regulation of peptidyl-serine phosphorylation
positive regulation of T cell proliferation
T cell activation
positive regulation of transcription from RNA polymerase II promoter
response to other organism
negative regulation of cell death
regulation of endoplasmic reticulum stress-induced eIF2 alpha phosphorylation
peptidyl-serine dephosphorylation
negative regulation of endoplasmic reticulum unfolded protein response
positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
positive regulation of cap-dependent translational initiation
positive regulation of cap-independent translational initiation
negative regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
protein kinase inhibitor activity
receptor binding
protein domain specific binding
receptor tyrosine kinase binding
eukaryotic initiation factor eIF2 binding
negative regulation of protein kinase activity
substrate-dependent cell migration, cell extension
actin filament organization
cell migration
lamellipodium assembly
positive regulation of actin filament polymerization
positive regulation of translation in response to stress
negative regulation of peptidyl-serine phosphorylation
positive regulation of T cell proliferation
T cell activation
positive regulation of transcription from RNA polymerase II promoter
response to other organism
negative regulation of cell death
regulation of endoplasmic reticulum stress-induced eIF2 alpha phosphorylation
peptidyl-serine dephosphorylation
negative regulation of endoplasmic reticulum unfolded protein response
positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
positive regulation of cap-dependent translational initiation
positive regulation of cap-independent translational initiation
negative regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
protein kinase inhibitor activity
receptor binding
protein domain specific binding
receptor tyrosine kinase binding
eukaryotic initiation factor eIF2 binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P717643 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644451 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389327 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249766 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912295 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722125 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916333 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238898 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643838 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741451 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728804 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607485 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153429 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858590 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956134 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854274 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80723 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150794 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138462 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P717643 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P717643 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P717643 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P717643 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 10469 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 47847 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10919550 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919549 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10919548 | - | - |
| Rat Genome 230 2.0 Array | 1373940_at | - | - |
| Rat Genome 230 2.0 Array | 1373940_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1373940_at | - | - |
| Affymetrix HT Rat Focus Array | 1373940_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800025817.rn.1 | TC0800003240.rn.1 | RTA TC |
| Rat Expression Set 230 | 1373940_PM_at | - | - |
| Rat Expression Set 230 | 1373940_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919548 | - | - |
| Rat Genome U34 Set | rc_AA955389_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358071 | - | - |

Nck1
Genomic Map