Assay Details
Gene Symbol: Mb21d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006243439.2 | XP_006243501.1 | 1 - 2 | 804 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
