Assay Details
| Gene Symbol: Ddx43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750545.3 | XP_003750593.1 | 15 - 16 | 2408 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Ddx43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750545.3 | XP_003750593.1 | 15 - 16 | 2408 | 83 | |
Set Membership: |
|