Assay Details
| Gene Symbol: Arpp19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031660.1 | NP_113848.1 | 2 - 3 | 166 | 80 | |
| XM_006243410.2 | XP_006243472.1 | 3 - 4 | 277 | 80 | ||
| GenBank mRNA | AJ005982.1 | - | 2 - 3 | 166 | 80 | |
| BC058461.1 | - | 3 - 4 | 212 | 6919944 | 80 | |
| FQ213926.1 | - | 1 - 2 | 247 | 80 | ||
| FQ221182.1 | - | 3 - 4 | 216 | 80 | ||
| FQ222843.1 | - | 3 - 4 | 215 | 80 | ||
| Gene Symbol: LOC100360828 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729972.4 | XP_002730018.2 | 1 | 203 | 80 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Arpp19
Genomic Map