Assay Details
| Gene Symbol: Rbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012640.2 | NP_036772.2 | 2 - 3 | 306 | 63 | |
| XM_006243607.1 | XP_006243669.1 | 3 - 4 | 486 | 63 | ||
| GenBank mRNA | CN542437.1 | - | 2 - 3 | 63 | ||
| M13949.1 | - | 2 - 3 | 306 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbp2
Genomic Map