Assay Details
| Gene Symbol: Mlip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008766417.1 | XP_008764639.1 | 9 - 10 | 2770 | 91 | |
| XM_008766418.1 | XP_008764640.1 | 8 - 9 | 2614 | 91 | ||
| XM_008766420.1 | XP_008764642.1 | 9 - 10 | 2578 | 91 | ||
| GenBank mRNA | BC092625.1 | - | 2190 | 91 | ||
| FQ224108.1 | - | 2433 | 91 | |||
More Information
Set Membership: |
|

Mlip
Genomic Map