Assay Details
| Gene Symbol: Zfp445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191802.1 | NP_001178731.1 | 5 - 6 | 966 | 98 | |
| XM_006244181.1 | XP_006244243.1 | 5 - 6 | 1040 | 98 | ||
| XM_006244183.2 | XP_006244245.1 | 5 - 6 | 967 | 98 | ||
| GenBank mRNA | FQ226616.1 | - | 4 - 5 | 713 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp445
Genomic Map