Assay Details
| Gene Symbol: Morf4l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011999.1 | NP_001011999.1 | 5 - 6 | 462 | 88 | |
| XM_008766445.1 | XP_008764667.1 | 6 - 7 | 545 | 88 | ||
| GenBank mRNA | BC078910.1 | - | 5 - 6 | 462 | 7131534 | 88 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Morf4l1
Genomic Map