Assay Details
| Gene Symbol: Nradd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139259.2 | NP_640352.1 | 2 - 3 | 221 | 79 | |
| GenBank mRNA | AF497263.1 | - | 2 - 3 | 225 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF497263.1, NM_139259.2 transcripts.
Set Membership: |
|

Nradd
Genomic Map