Assay Details
| Gene Symbol: Nradd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006243921.2 | XP_006243983.1 | 2 - 3 | 371 | 75 | |
| GenBank mRNA | AF497264.1 | - | 2 - 3 | 219 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF497264.1, XM_006243921.2 transcripts.
Set Membership: |
|

Nradd
Genomic Map