Assay Details
| Gene Symbol: Panx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270548.1 | NP_001257477.1 | 1 - 2 | 620 | 69 | |
| NM_001270549.1 | NP_001257478.1 | 1 - 2 | 620 | 69 | ||
| NM_199397.2 | NP_955429.1 | 1 - 2 | 620 | 69 | ||
| GenBank mRNA | AJ557015.1 | - | 1 - 2 | 620 | 69 | |
| GQ499839.1 | - | 1 - 2 | 284 | 69 | ||
| GQ499840.1 | - | 1 - 2 | 284 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Panx1
Genomic Map