Assay Details
| Gene Symbol: Zfp653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106807.1 | NP_001100277.1 | 2 - 3 | 345 | 79 | |
| XM_006242635.2 | XP_006242697.1 | 2 - 3 | 642 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp653
Genomic Map