Assay Details
| Gene Symbol: RGD1309534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014206.1 | NP_001014228.1 | 1 - 2 | 33 | 56 | |
| GenBank mRNA | BC085915.1 | - | 1 - 2 | 33 | 7107444 | 56 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC085915.1, NM_001014206.1 transcripts.
Set Membership: |
|

RGD1309534
Genomic Map