Assay Details
Gene Symbol: Zfp810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | U78134.1 | - | 1 | 57 | 72 |
Gene Symbol: Zfp717 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766011.1 | XP_008764233.1 | 12 | 1940 | 72 | |
XM_008766012.1 | XP_008764234.1 | 12 | 1782 | 72 | ||
XM_008766013.1 | XP_008764235.1 | 11 | 1889 | 72 | ||
XM_008766014.1 | XP_008764236.1 | 11 | 1731 | 72 | ||
XM_008766015.1 | XP_008764237.1 | 12 | 1737 | 72 | ||
XM_008766016.1 | XP_008764238.1 | 11 | 1876 | 72 | ||
XM_008766017.1 | XP_008764239.1 | 11 | 1718 | 72 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
