Assay Details
| Gene Symbol: Tmeff1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023020.2 | NP_075409.1 | 3 - 4 | 724 | 62 | |
| GenBank mRNA | AJ250730.1 | - | 3 - 4 | 521 | 62 | |
| BC129093.1 | - | 3 - 4 | 724 | 8363015 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmeff1
Genomic Map