Assay Details
| Gene Symbol: Nr4a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031628.1 | NP_113816.1 | 3 - 4 | 1659 | 94 | |
| XM_008763702.1 | XP_008761924.1 | 4 - 5 | 3453 | 94 | ||
| XM_008763703.1 | XP_008761925.1 | 3 - 4 | 3381 | 94 | ||
| GenBank mRNA | D38530.1 | - | 3 - 4 | 1659 | 94 | |
| DQ268830.1 | - | 3 - 4 | 1659 | 94 | ||
| X86003.1 | - | 1 - 2 | 1446 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nr4a3
Genomic Map