Assay Details
| Gene Symbol: LOC103690035 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006238153.2 | XP_006238215.2 | 4 - 5 | 2325 | 124 | |
| XM_008763706.1 | XP_008761928.1 | 4 - 5 | 2337 | 124 | ||
| Gene Symbol: Tgfbr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012775.2 | NP_036907.2 | 1040 | 124 | ||
| XM_006238030.2 | XP_006238092.2 | 1054 | 124 | |||
| GenBank mRNA | BC087035.1 | - | 982 | 7098473 | 124 | |
| L26110.1 | - | 1009 | 124 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103690035
Genomic Map