Assay Details
| Gene Symbol: Sec61b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106654.2 | NP_001100124.1 | 1 - 2 | 102 | 97 | |
| GenBank mRNA | FQ210322.1 | - | 1 - 2 | 90 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001106654.2 transcripts.
Set Membership: |
|

Sec61b
Genomic Map