Assay Details
| Gene Symbol: Sec61b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106654.2 | NP_001100124.1 | 2 - 3 | 207 | 139 | |
| GenBank mRNA | BC167110.1 | - | 2 - 3 | 207 | 6887572 | 139 |
| FQ210322.1 | - | 2 - 3 | 195 | 139 | ||
| FQ217838.1 | - | 2 - 3 | 160 | 139 | ||
| FQ221073.1 | - | 2 - 3 | 154 | 139 | ||
| FQ221908.1 | - | 2 - 3 | 151 | 139 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sec61b
Genomic Map