Assay Details
| Gene Symbol: Ctsq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139262.1 | NP_640355.1 | 4 - 5 | 409 | 124 | |
| GenBank mRNA | AF187323.1 | - | 4 - 5 | 409 | 124 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ctsq
Genomic Map