Assay Details
| Gene Symbol: F12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014006.1 | NP_001014028.1 | 12 - 13 | 1700 | 86 | |
| GenBank mRNA | BC088187.1 | - | 12 - 13 | 1700 | 7370526 | 86 |
More Information
Set Membership: |
|

F12
Genomic Map