Assay Details
| Gene Symbol: Cpa6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107900.1 | NP_001101370.1 | 4 - 5 | 448 | 74 | |
| XM_008763496.1 | XP_008761718.1 | 4 - 5 | 512 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P372998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18215 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241898 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684103 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307381 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333246 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65709 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926902 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124381 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152423 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158056 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93401 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535140 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563154 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503784 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967586 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832520 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816185 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43170 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269318 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261218 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834080 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711630 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169195 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146876 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P372998 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P372998 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P372998 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P105462 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P372998 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 80071 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 96622 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 91720 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10867244 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10867251 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10867252 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10867249 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10867253 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10867248 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10867246 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10867247 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10867245 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10867241 | - | - |
| Rat Genome 230 2.0 Array | 1384454_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1384454_at | - | - |
| Affymetrix HT Rat Focus Array | 1384454_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1384454_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500000437.rn.1 | TC0500000067.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500000439.rn.1 | TC0500000067.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500000432.rn.1 | TC0500000067.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500000435.rn.1 | TC0500000067.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500000440.rn.1 | TC0500000067.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500000433.rn.1 | TC0500000067.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500000434.rn.1 | TC0500000067.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500000436.rn.1 | TC0500000067.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500000442.rn.1 | TC0500000067.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500000444.rn.1 | TC0500000067.rn.1 | RTA TC |
| Rat Expression Set 230 | 1384454_at | - | - |
| Rat Expression Set 230 | 1384454_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10867241 | - | - |
| Rat Genome U34 Set | rc_AA819425_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20712006 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364825 | - | - |

Cpa6
Genomic Map