Assay Details
| Gene Symbol: Jph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106630.1 | NP_001100100.1 | 2 - 3 | 1335 | 68 | |
| XM_006237719.2 | XP_006237781.1 | 2 - 3 | 1335 | 68 | ||
| XM_006237720.2 | XP_006237782.1 | 2 - 3 | 1335 | 68 | ||
| XM_006237721.2 | XP_006237783.1 | 2 - 3 | 1335 | 68 | ||
| XM_006237722.2 | XP_006237784.1 | 2 - 3 | 1335 | 68 | ||
| XM_006237723.2 | XP_006237785.1 | 2 - 3 | 1335 | 68 | ||
| XM_008763475.1 | XP_008761697.1 | 2 - 3 | 1335 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P076555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53386 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19810 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641302 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59586 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737083 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469569 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845739 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545933 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585898 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175261 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283384 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816643 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423726 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280208 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316520 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381140 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662639 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525703 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71526 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87777 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116540 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24042 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3843 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P076555 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P076555 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P076555 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P23135 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 89917 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 103402 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 64533 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 75675 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10867085 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10867082 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10867084 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10867083 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10867086 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10867081 | - | - |
| Rat Genome 230 2.0 Array | 1376015_at | - | - |
| Rat Genome 230 2.0 Array | 1376015_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500000030.rn.1 | TC0500000014.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500000029.rn.1 | TC0500000014.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500000031.rn.1 | TC0500000014.rn.1 | RTA TC |
| Rat Expression Set 230 | 1376015_at | - | - |
| Rat Expression Set 230 | 1376015_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10867081 | - | - |
| Rat Genome U34 Set | rc_AI030414_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21719823 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358279 | - | - |

Jph1
Genomic Map