Assay Details
| Gene Symbol: Trmt44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130061.1 | NP_001123533.1 | 10 - 11 | 1902 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P524851 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P068344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878456 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597982 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126331 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P068344 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P524851 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P524851 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P068344 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P524851 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P068344 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P21989 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10773429 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10773426 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10773428 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10773425 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10773432 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10773431 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10773430 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10773433 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10773427 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10773434 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10773424 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10773423 | - | - |
| Rat Genome 230 2.0 Array | 1390222_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1390222_at | - | - |
| Affymetrix HT Rat Focus Array | 1390222_at | - | - |
| Affymetrix HT Rat Focus Array | 1390222_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1400005612.rn.1 | TC1400000807.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400005611.rn.1 | TC1400000807.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400005613.rn.1 | TC1400000807.rn.1 | RTA TC |
| Rat Expression Set 230 | 1390222_PM_at | - | - |
| Rat Expression Set 230 | 1390222_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10773423 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22232095 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1366679 | - | - |

Trmt44
Genomic Map