Assay Details
| Gene Symbol: LOC100912481 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749683.2 | XP_003749731.1 | 13 - 14 | 1458 | 131 | |
| XM_003749684.2 | XP_003749732.1 | 14 - 15 | 1473 | 131 | ||
| XM_006236016.1 | XP_006236078.1 | 14 - 15 | 1476 | 131 | ||
| XM_006236017.1 | XP_006236079.1 | 13 - 14 | 1455 | 131 | ||
| Gene Symbol: Ewsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025632.2 | NP_001020803.2 | 13 - 14 | 1453 | 131 | |
| XM_006251201.2 | XP_006251263.2 | 13 - 14 | 1438 | 131 | ||
| XM_006251202.2 | XP_006251264.2 | 13 - 14 | 1435 | 131 | ||
| XM_006251203.2 | XP_006251265.2 | 13 - 14 | 1464 | 131 | ||
| GenBank mRNA | BC098822.1 | - | 4 - 5 | 487 | 131 | |
| BC162004.1 | - | 13 - 14 | 1453 | 9089248 | 131 | |
| FQ221459.1 | - | 4 - 5 | 600 | 131 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100912481
Genomic Map